Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MTAVGRRCPA LGSRGAAGEP EAGSDYVKFS KEKYILDSSP EKLHKELEEE LKLSSTDLRS HAWYHGRIPR EVSETLVQRN GDFLIRDSLT SLGDYVLTCR WRNQALHFKI
Eph receptor-binding protein which may be a positive regulator of TCR signaling. Binding to BCAR1 is required to induce membrane ruffling and promote EGF-dependent cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cas/HEF1-associated signal transducer; Chat-H; Novel SH2-containing protein 3; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; SHEP1
Gene Aliases: CHAT; NSP3; PRO34088; SH2D3C; SHEP1; UNQ272/PRO309/PRO34088
UniProt ID: (Human) Q8N5H7
Entrez Gene ID: (Human) 10044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support