Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MNIMDFNVKK LAADAGTFLS RAVQFTEEKL GQAEKTELDA HLENLLSKAE CTKIWTEKIM KQTEVLLQPN PNARIEEFVY EKLDRKAPSR INNPELLGQY MIDAGTEFGP GTAYGNALIK CGETQKRIGT ADRELIQTSA LNFLTPLRNF IEGDYKTIAK ERKLLQNKRL DLDAAKTRLK KAKAAETRNS SEQELRITQS EFDRQAEITR LLLEGISSTH AHHLRCLNDF VEAQMTYYAQ CYQYMLDLQK QLGSFPSNYL SNNNQTSVTP VPSVLPNAIG SSAMASTSGL VITSPSNLSD LKECSGSRKA RVLYDYDAAN STELSLLADE VITVFSVVGM DSDWLMGERG NQKGKVPITY LELLN
Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bax-interacting factor 1; Bif-1; BIF1; endophilin b1; Endophilin-B1; protein phosphatase 1, regulatory subunit 70; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like endophilin B1; testicular tissue protein Li 172
Gene Aliases: Bif-1; CGI-61; dJ612B15.2; KIAA0491; PPP1R70; SH3GLB1
UniProt ID: (Human) Q9Y371
Entrez Gene ID: (Human) 51100
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support