Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is A549 cell lysate.
PA5-20817 can be used with blocking peptide PEP-0931.
Upon entry into mitosis, the cell's microtubule (MT) network forms the mitotic spindle, allowing the segregation of paired chromosomes. Proteinaceous structures on centromeric chromatin termed kinetochores (KT) are essential for the proper attachment of the chromosomes to the spindle MTs. A recently discovered spindle and kinetochore complex, comprised of proteins SKA1, SKA2, and SKA3, has been found to be required for stable KT-MT interactions and timely anaphase onset. Depletion of either SKA1 or SKA2 by siRNA results in the loss of both proteins from the KT, but does not impact overall KT structure. Cells depleted of the SKA complex undergo a prolonged checkpoint-dependent delay in a metaphase-like state, indicating the importance of the SKA complex in the maintenance of the metaphase plate and spindle checkpoint silencing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SKA complex subunit 1; Spindle and kinetochore-associated protein 1; spindle and KT (kinetochore) associated 1
Gene Aliases: 2810433K01Rik; AV117428; C18orf24; RGD1310784; SKA1
UniProt ID: (Human) Q96BD8, (Rat) B0BN28, (Mouse) Q9CPV1
Entrez Gene ID: (Human) 220134, (Rat) 291441, (Mouse) 66468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support