Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This target displays homology in the following species: Cow: 90%; Dog: 91%; Horse: 85%; Human: 100%; Mouse: 79%; Rat: 79%
SLC13A4 gene ontology annotations related to this gene include sodium ion transport; transmembrane transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/sulfate cotransporter SUT-1; NaS2; solute carrier family 13 (sodium/sulfate symporter), member 4; solute carrier family 13 (sodium/sulfate symporters), member 4; solute carrier family 13 (sodium/sulphate symporters), member 4; Solute carrier family 13 member 4; sulphate transporter 1
Gene Aliases: NAS2; SLC13A4; SUT-1; SUT1
UniProt ID: (Human) Q9UKG4
Entrez Gene ID: (Human) 26266
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support