Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SLC17A1.
SLC17A1 (Solute Carrier Family 17 Member 1) is a Protein Coding gene. Diseases associated with SLC17A1 include Gout and Hyperuricemia. Among its related pathways are Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds and Uricosurics Pathway, Pharmacodynamics. Gene Ontology (GO) annotations related to this gene include symporter activity and phosphate ion transmembrane transporter activity. An important paralog of this gene is SLC17A2. Also, it is involved in cotransport of sodium and phosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/PI cotransporter 1; Renal Na(+)-dependent phosphate cotransporter 1; Renal sodium-dependent phosphate transport protein 1; Renal sodium-phosphate transport protein 1; Sodium-dependent phosphate transport protein 1; sodium-dependent phosphate/anion cotransporter; Sodium/phosphate cotransporter 1; sodium/phosphate type I cotransporter; solute carrier family 17 (sodium/hydrogen exchanger), member 1; Solute carrier family 17 member 1; solute carrier family 17 vesicular glutamate transporter), member 1
Gene Aliases: NAPI-1; Napi1; Npt1; Slc17a1
UniProt ID: (Mouse) Q61983
Entrez Gene ID: (Mouse) 20504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support