Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TACVLTGQP FDTIKVKMQT FPDLYKGLTD CFLKTYAQVG LRGFYKGTGP ALMAYVAENS VLFMCYGFCQ QFVRKVAGMD KQ (21-101 aa encoded by BC100935)
Located between the protocadherin beta and gamma gene clusters on chromosome 5, this intronless gene encodes a protein that is highly similar to an ornithine transporter localized in the mitochondrial inner membrane. The encoded protein most likely plays a role in metabolism as a mitochondrial transport protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial ornithine transporter; Mitochondrial ornithine transporter 2; ornithine transporter 2; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 2; Solute carrier family 25 member 2
Gene Aliases: ORC2; ORNT2; SLC25A2
UniProt ID: (Human) Q9BXI2
Entrez Gene ID: (Human) 83884, (Mouse) 83885, (Rat) 291640
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support