Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLCLCLYVP VIGEAQTEFQ YFESKGLPAE LKSIFKLSVF IPSQEFSTYR QWKQ (1-53 aa encoded by BC103933)
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-Mg/Pi carrier; calcium-binding mitochondrial carrier protein SCaMC-2; Mitochondrial adenyl nucleotide antiporter SLC25A25; mitochondrial ATP-Mg/Pi carrier protein; Mitochondrial ATP-Mg/Pi carrier protein 3; Mitochondrial Ca(2+)-dependent solute carrier protein 3; mitochondrial Ca2+-dependent solute carrier; Mitochondrial calcium-dependent solute carrier; peroxisomal Ca(2+)-dependent solute carrier-like protein; peroxisomal Ca-dependent solute carrier-like protein; SCaMC-2; short calcium-binding mitochondrial carrier 2; Short calcium-binding mitochondrial carrier protein 2; small calcium-binding mitochondrial carrier 2; small calcium-binding mitochondrial carrier protein 2; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25; Solute carrier family 25 member 25
Gene Aliases: 1110030N17Rik; APC3; KIAA1896; MCSC; MCSC3; mKIAA1896; PCSCL; SCAMC-2; SCAMC2; SLC25A25; UNQ549/PRO1106
UniProt ID: (Human) Q6KCM7, (Mouse) A2ASZ8, (Rat) Q8K3P6
Entrez Gene ID: (Human) 114789, (Mouse) 227731, (Rat) 246771
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support