Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: mouse heart, mouse brain
Immunogen sequence: EILSRALEVS PPRCLVLECT HVCSIDYTVV LGLGELLQDF QKQGVALAFV GLQVPVLRVL LSADLKGFQY FSTLEEAEKH LRQEPGTQPY NIREDSILDQ K
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anion exchanger Slc26a11; KBAT; Kidney brain anion transporter; novel sulfate transporter protein (Slc26a11); slc26a11; Sodium-independent sulfate anion transporter; Solute carrier family 26 member 11
Gene Aliases: F630021I08Rik; Slc26a11
UniProt ID: (Mouse) Q80ZD3
Entrez Gene ID: (Mouse) 268512
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support