Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSRLPTYYL TTFFVGEGLS GLLPALVALA QGSGLTTCVN VTEISDSVPS PVPTRETDIA QGVPRALVSA LPGMEAPLSH LESRYLPAHF SPLVFFLLLS IMMACCLVAF FVLQRQPRCW EASVEDLLND QVTLHSIRLR EENDLGPAGM VDSSQGQGYL EEKAAPCCPA HLAFV (130-303 aa encoded by BC009750)
This gene encodes a riboflavin transporter protein that is strongly expressed in the intestine and likely plays a role in intestinal absorption of riboflavin. The protein is predicted to have eleven transmembrane domains and a cell surface localization signal in the C-terminus. Mutations at this locus have been associated with Brown-Vialetto-Van Laere syndrome and Fazio-Londe disease. [provided by RefSeq, Mar 2012].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ct054; hRFT2; RFT2; Riboflavin transporter 2; solute carrier family 52 (riboflavin transporter), member 3; Solute carrier family 52, riboflavin transporter, member 3
Gene Aliases: 2310046K01Rik; bA371L19.1; BVVLS; BVVLS1; C20orf54; hRFT2; RFT2; RFVT3; SLC52A3
UniProt ID: (Human) Q9NQ40, (Mouse) Q9D6X5
Entrez Gene ID: (Human) 113278, (Mouse) 69698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support