Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: U-87MG, HT-29, 293T, Mouse brain, Rat brain.
Immunogen sequence: PPTKEQIRTT TFWSKKNLVV KENCSPKEEP YKMQEKSILR CSENNETINH IIPNGKSEDS IKGLQPEDVN LLVTCREEGN PVASLGHSEA ETPVDAYSNG QAALMGEKER KKETDDGGRY WKFIDWFCGF KSKSLSKRSL RDLMEEEAVC LQMLEETRQV K
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCW2 protein; Na(+)/myo-inositol cotransporter; SMIT1; sodium myo-inositol transporter 1; Sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; Sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; Solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3
Gene Aliases: AA623876; BCW2; BF642829; SLC5A3; SMIT; SMIT1; SMIT2
UniProt ID: (Human) P53794, (Mouse) Q9JKZ2
Entrez Gene ID: (Human) 6526, (Mouse) 53881, (Rat) 114507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support