Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KATFNYENCL KKVSLLLTNT FDLEDGFLTA SNLEQVKGYL ASAYPSKYSE MFPQIKNCSL ESELDTAVQ
Mediates the calcium-dependent uptake of imino acids such as L-proline, N-methyl-L-proline and pipecolate as well as N-methylated amino acids. Involved in the transport of glycine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurotransmitter transporter RB21A; orphan transporter XT3; Sodium- and chloride-dependent transporter XTRP3; Sodium/imino-acid transporter 1; solute carrier family 6 (neurotransmitter transporter), member 20; solute carrier family 6 (proline IMINO transporter), member 20; Solute carrier family 6 member 20; Transporter rB21A homolog; X transporter protein 3
Gene Aliases: SIT1; SLC6A20; XT3; XTRP3
UniProt ID: (Human) Q9NP91
Entrez Gene ID: (Human) 54716
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support