Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
SLX4IP/C20orf94 interacts with SLX4/BTBD12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SLX4IP; SLX4-interacting protein; UPF0492 protein C20orf94 homolog
Gene Aliases: 2210009G21Rik; 2410004I22Rik; bA204H22.1; bA254M13.1; C20orf94; C85661; dJ1099D15.3; SLX4IP; Slxip
UniProt ID: (Human) Q5VYV7, (Mouse) Q9D7Y9
Entrez Gene ID: (Human) 128710, (Mouse) 74243
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support