Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SMARCA5 Antibodies

          Abnova

          SMARCA5 Polyclonal Antibody, MaxPab™

          View all (15) SMARCA5 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SMARCA5 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          SMARCA5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SMARCA5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SMARCA5 Antibody (H00008467-B01P) in WB

          Western Blot analysis of SMARCA5 expression in transfected 293T cell line by SMARCA5 MaxPab polyclonal antibody. Lane 1: SMARCA5 transfected lysate(115.72 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SMARCA5 Antibody in Western Blot (WB)

          Product Details

          H00008467-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SMARCA5 (AAH23144.1, 1 a.a. approximately 1052 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSSAAEPPPP PPPESAPSKP AASIASGGSN SSNKGGPEGV AAQAVASAAS AGPADAEMEE IFDDASPGKQ KEIQEPDPTY EEKMQTDRAN RFEYLLKQTE LFAHFIQPAA QKTPTSPLKM KPGRPRIKKD EKQNLLSVGD YRHRRTEQEE DEELLTESSK ATNVCTRFED SPSYVKWGKL RDYQVRGLNW LISLYENGIN GILADEMGLG KTLQTISLLG YMKHYRNIPG PHMVLVPKST LHNWMSEFKR WVPTLRSVCL IGDKEQRAAF VRDVLLPGEW DVCVTSYEML IKEKSVFKKF NWRYLVIDEA HRIKNEKSKL SEIVREFKTT NRLLLTGTPL QNNLHELWSL LNFLLPDVFN SADDFDSWFD TNNCLGDQKL VERLHMVLRP FLLRRIKADV EKSLPPKKEV KIYVGLSKMQ REWYTRILMK DIDILNSAGK MDKMRLLNIL MQLRKCCNHP YLFDGAEPGP PYTTDMHLVT NSGKMVVLDK LLPKLKEQGS RVLIFSQMTR VLDILEDYCM WRNYEYCRLD GQTPHDERQD SINAYNEPNS TKFVFMLSTR AGGLGINLAT ADVVILYDSD WNPQVDLQAM DRAHRIGQTK TVRVFRFITD NTVEERIVER AEMKLRLDSI VIQQGRLVDQ NLNKIGKDEM LQMIRHGATH VFASKESEIT DEDIDGILER GAKKTAEMNE KLSKMGESSL RNFTMDTESS VYNFEGEDYR EKQKIAFTEW IEPPKRERKA NYAVDAYFRE ALRVSEPKAP KAPRPPKQPN VQDFQFFPPR LFELLEKEIL FYRKTIGYKV PRNPELPNAA QAQKEEQLKI DEAESLNDEE LEEKEKLLTQ GFTNWNKRDF NQFIKANEKW GRDDIENIAR EVEGKTPEEV IEYSAVFWER CNELQDIEKI MAQIERGEAR IQRRISIKKA LDTKIGRYKA PFHQLRISYG TNKGKNYTEE EDRFLICMLH KLGFDKENVY DELRQCIRNS PQFRFDWFLK SRTAMELQRR CNTLITLIER ENMELEEKEK AEKKKRGPKP STQKRKMDGA PDGRGRKKKL KL

          Target Information

          SMARCA5 is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. SMARCA5 is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein encoded by this gene is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The encoded protein is similar in sequence to the Drosophila ISWI chromatin remodeling protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hSNF2H; Sucrose nonfermenting protein 2 homolog; sucrose nonfermenting-like 5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5

          View more View less

          Gene Aliases: hISWI; hSNF2H; ISWI; SMARCA5; SNF2H; WCRF135

          View more View less

          UniProt ID: (Human) O60264

          View more View less

          Entrez Gene ID: (Human) 8467

          View more View less

          Function(s)
          DNA binding helicase activity protein binding ATP binding ATPase activity nucleosome binding histone binding
          Process(es)
          chromatin silencing at rDNA double-strand break repair nucleosome assembly chromatin remodeling DNA-templated transcription, initiation regulation of transcription from RNA polymerase II promoter covalent chromatin modification nucleosome positioning CENP-A containing nucleosome assembly ATP-dependent chromatin remodeling positive regulation of gene expression, epigenetic positive regulation of transcription, DNA-templated
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0