Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DLKLWEIGK IIGGMWRDLT DEEKQEYLNE YEAEKIEYNE SMKAYHNSPA YLAYINAKSR AEAALEEESR QRQSRMEKGE PYMSIQPAED PDDYDDGFSM KHTATARFQR NHRLISEILS ESVVPDVRSV VTTARMQVLK RQVQSLMVHQ RKLEAELLQI EERHQEKKRK FLESTDSFNN ELKRLCGLKV EVDMEKIAAE IAQAEEQARK RQEEREKEAA EQAERSQSSI VPEEEQAANK GEEKKDDENI PMETEETHLE ETTESQQNGE EGTSTPEDKE SGQEGVDSMA EEGTSDSNTG SESNSATVEE PPTDPIPEDE KKE (90-411 aa encoded by BC007082)
The protein encoded by this gene is part of the large ATP-dependent chromatin remodeling complex SWI/SNF, which is required for transcriptional activation of genes normally repressed by chromatin. The encoded protein, either alone or when in the SWI/SNF complex, can bind to 4-way junction DNA, which is thought to mimic the topology of DNA as it enters or exits the nucleosome. The protein contains a DNA-binding HMG domain, but disruption of this domain does not abolish the DNA-binding or nucleosome-displacement activities of the SWI/SNF complex. Unlike most of the SWI/SNF complex proteins, this protein has no yeast counterpart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAF57; BRG1-associated factor 57; chromatin remodeling complex BRG1-associated factor 57; SMARCE 1; SWI/SNF-related matrix-associated actin-dependent regulator chromatin subfamily E member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin e1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1
Gene Aliases: 2810417B20Rik; 5830412H02Rik; 9030408N19Rik; Baf27; BAF57; CSS5; RGD1304726; SMARCE1
UniProt ID: (Human) Q969G3, (Rat) Q56A18, (Mouse) O54941
Entrez Gene ID: (Human) 6605, (Rat) 303518, (Mouse) 57376
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support