Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Plays essential roles in both eye and limb development. Probable regulator of osteoblast differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: secreted modular calcium binding protein 1; Secreted modular calcium-binding protein 1; SMOC-1; SPARC-related gene protein; SPARC-related modular calcium-binding protein 1; SPARC-related protein
Gene Aliases: 2600002F22Rik; AI848508; OAS; SMOC1; SRG
UniProt ID: (Human) Q9H4F8, (Mouse) Q8BLY1
Entrez Gene ID: (Human) 64093, (Mouse) 64075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support