Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MNMSKQPVS NVRAIQANIN IPMGAFRPGA GQPPRRKECT PEVEEGVPPT SDEEKKPIPG AKKLPGPAVN LSEIQNIKSE LKYVPKAEQ (1-88 aa encoded by BC005948)
SMPX plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deafness, X-linked 6, sensorineural; Small muscular protein; Stretch-responsive skeletal muscle protein
Gene Aliases: DFN6; DFNX4; SMPX; SRMX
UniProt ID: (Human) Q9UHP9
Entrez Gene ID: (Human) 23676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support