Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRCSQCRVA KYCSAKCQKK AWPDHKRECK CLKSCKPRYP PDSVRLLGRV VFKLMDGAPS ESEKLYSFYD LESNINKLTE DRKEGLRQLV MTFQHFMREE IQDASQLPPA FDLFEAFAKV ICNSFTICNA EMQEVGVGLY PSISLLNHSC DPNCSIVFNG PHLLLRAVRD IEVGEELTIC YLDMLMTSEE RRKQLRDQYC FECDCFRCQT QDKDADMLTG DEQVWKEVQE SLKKIEELKA HWKWEQVLAM CQAIISSNSE RLPDINIYQL KVLDCAMDAC INLGLLEEAL FYGTRTMEPY RIFFPGSHPV RGVQVMKVGK LQLHQGMFPQ AMKNLRLAFD IMRVTHGREH SLIEDLILLL EECDANIRAS (60-428 aa encoded by BC031010)
SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA74P14.1 (novel protein); Histone-lysine N-methyltransferase SMYD3; SET and MYND domain-containing protein 3; Zinc finger MYND domain-containing protein 1; zinc finger protein, subfamily 3A (MYND domain containing), 1; zinc finger, MYND domain containing 1
Gene Aliases: bA74P14.1; KMT3E; SMYD3; ZMYND1; ZNFN3A1
UniProt ID: (Human) Q9H7B4
Entrez Gene ID: (Human) 64754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support