Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SNF1 Antibodies

          Invitrogen

          SNF1 Polyclonal Antibody

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SNF1 Polyclonal Antibody

          • Antibody Testing Data (1)
          SNF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SNF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SNF1 Antibody (PA5-144419) in WB

          Western blot analysis of SNF1 in recombinant protein using a dilution of 1:2,000. Samples were incubated with SNF1 polyclonal antibody (Product # PA5-144419). Secondary: Goat polyclonal to rabbit IgG at 1:50,000 dilution. Predicted band size: 27 kDa. Observed band size: 36 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SNF1 Antibody in Western Blot (WB)

          Product Details

          PA5-144419

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Yeast

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) SNF1 protein (435-633aa)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          3.91 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.03% Proclin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3091714

          Target Information

          Serine/threonine protein kinase essential for release from glucose repression (PubMed:3526554, PubMed:6366512, PubMed:3049551, PubMed:1944227, PubMed:8289797, PubMed:8628258, PubMed:25869125). Catalytic subunit of the AMP-activated protein kinase complex also known as the SNF1 kinase complex (Snf1c), a central regulator of cellular energy homeostasis, which, in response to a fall in intracellular ATP levels, activates energy-producing pathways and inhibits energy-consuming processes (PubMed:8289797, PubMed:26667037). The complex phosphorylates histone H3 to form H3S10ph, which promotes H3K14ac formation, leading to transcriptional activation through TBP recruitment to the promoters (PubMed:15719021). The complex also negatively regulates the HOG1 MAPK pathway in ER stress response including unfolded protein response (UPR) (PubMed:25730376, PubMed:26394309). Under nutrient/energy depletion, the complex phosphorylates and activates PAS kinase PSK1 which in turn activates PBS1, leading to the inhibition of the TORC1 signaling pathway (PubMed:25428989). SNF1 also interacts and phosphorylates adenylate cyclase CYR1 and negatively regulates the protein kinase A signaling pathway (PubMed:26309257). Also phosphorylates and regulates the transcriptional activator CAT8 (PubMed:15121831).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Carbon catabolite-derepressing protein kinase; CAT1; CCR1; GLC2; PAS14; Snf1p; Sucrose nonfermentating protein 1

          View more View less

          Gene Aliases: CAT1; CCR1; D8035.20; GLC2; HAF3; PAS14; SNF1; YDR477W

          View more View less

          UniProt ID: (Baker's yeast) P06782

          View more View less

          Entrez Gene ID: (Baker's yeast) 852088

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity AMP-activated protein kinase activity ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          replicative cell aging invasive growth in response to glucose limitation carbohydrate metabolic process regulation of carbohydrate metabolic process protein phosphorylation cellular response to nitrogen starvation pseudohyphal growth cell adhesion phosphorylation negative regulation of translation biofilm formation positive regulation of gluconeogenesis positive regulation of filamentous growth of a population of unicellular organisms in response to starvation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0