Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GSAYVAYNIH VNGVLHCRVR YSQLLGLHEQ LRKEYGANVL PAFPPKKLFS LTPAEVEQRR EQLEKYMQAV RQDPLLGSSE TFNSFLRRAQ QETQQVPTEE VSLEVLLSNG QKVLVNVLTS DQTEDVLEAV AAKLDLPDDL IGYFSLFLVR EKEDGAFSFV RKLQEFELPY VSVTSLRSQE YKIVLRKSYW DSAYDDDVME NRVGLNLLYA
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a B41 domain. This protein interacts with the cytoplasmic domain of P-selectin, and may function in the intracellular trafficking of P-selectin. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sorting nexin-17
Gene Aliases: KIAA0064; SNX17
UniProt ID: (Human) Q15036, (Rat) Q6AYS6
Entrez Gene ID: (Human) 9784, (Rat) 298836
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support