Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FPHDQRTLA AAAAAQQGFL FPPGITYKPG DNYPVQFIPS TMAAAAASGL SPLQLQKGHV SHPQINQRLK GLSDRFGRNL DTFEHGGGHS YNHKQIEQLY AAQLASMQVS PGAKMPSTPQ PPNTAGTVSP TGIKNEKRGT SPVTQVKDEA AAQPLNLSSR PKTAEPVKSP TSPTQNLFPA SKTSPVNLPN KSSIPSPIGG SLGRGSSLDI LSSLNSPALF GDQDTVMKAI QEARKMREQI QREQQQQQPH GVDGKLSSIN NMGLNSCRNE KERTRFENLG PQLTGKSNED GKLGPGVIDL TRPEDAEGSK AMNGSAAKLQ QYYCWPTGGA TVAEARVYRD ARGRASSEPH IKRPMNAFMV WAKDERRKIL QAFPDMHNSN ISKILGSRWK SMSNQEKQPY YEEQARLSKI HLEKYPNYKY KPRPKRTCIV DGKKLRIGEY KQLMRSRRQE MRQFFTVGQQ PQIPITTGTG VVYPGAITMA TTTPSPQMTS DCSSTSASPE PSLPVIQSTY GMKTDGGSLA GNEMINGEDE MEMYDDYEDD PKSDYSSENE APEAVSAN (272-828 aa encoded by BC047064)
Transcriptional activator. Binds specifically to the DNA sequence 5'-AACAAT-3'. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box containing gene 6; Transcription factor SOX-6
Gene Aliases: AI987981; HSSOX6; Sox-6; SOX-LZ; SOX6; SOXD
UniProt ID: (Human) P35712, (Rat) A0A0G2JTZ2, (Mouse) P40645
Entrez Gene ID: (Human) 55553, (Rat) 293165, (Mouse) 20679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support