Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Projection protein PF6 homolog; sperm associated antigen 17; sperm associated antigen 17, pseudogene; Sperm-associated antigen 17
Gene Aliases: 4931427F14Rik; CT143; PF6; SPAG17; Spag17-ps
UniProt ID: (Human) Q6Q759, (Mouse) Q5S003
Entrez Gene ID: (Human) 200162, (Mouse) 74362, (Rat) 690746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support