Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: human U-87MG whole cell, human SHG-44 whole cell, human MDA-MB-231 whole cell, human K562 whole cell, rat C6 whole cell, mouse smooth muscle tissue. IHC: human colon cancer tissue, mouse brain tissue.
This is a secreted protein. Found in the extracellular matrix. It's highly expressed in lymph node, brain, heart, lung, skeletal muscle, ovary, small intestine, and colon, with lower levels in placenta, pancreas, testis, spleen, and thymus, and no expression in kidney, liver, and peripheral blood leukocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Extracellular matrix protein 2; Hevin; High endothelial venule protein; MAST 9; Matrix glycoprotein Sc1; proliferation-inducing protein 33; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1
Gene Aliases: Ecm2; hevin; MAST 9; MAST9; PIG33; SC1; SPARCL1
UniProt ID: (Human) Q14515, (Mouse) P70663, (Rat) P24054
Entrez Gene ID: (Human) 8404, (Mouse) 13602, (Rat) 25434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support