Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MAVLLLLLRA LRRGPGPGPR PLWGPGPAWS PGFPARPGRG RPYMASRPPG DLAEAGGRAL QSLQLRLLTP TFEGINGLLL KQHLVQNPVR LWQLLGGTFY FNTSRLKQKN KEKDKSKGKA PEEDEEERRR RERDDQMYRE RLRTLLVIAV VMSLLNALST SGGSISWNDF VHEMLAKGEV QRVQVVPESD VVEVYLHPGA VVFGRPRLAL MYRMQVANID KFEEKLRAAE DELNIEAKDR IPVSYKRTGF FGNALYSVGM TAVGLAILWY VFRLAGMTGR EGGFSAFNQL KMARFTIVDG KMGKGVSFKD VAGMHEAKLE VREFVDYLKS PERFLQLGAK VPKGALLLGP PGCGKTLLAK AVATEAQVPF LAMAGPEFVE VIGGLGAARV RSLFKEARAR APCIVYIDEI DAVGKKRSTT MSGFSNTEEE QTLNQLLVEM DGMGTTDHVI VLASTNRADI LDGALMRPGR LDRHVFIDLP TLQERREIFE QHLKSLKLTQ SSTFYSQRLA ELTPGFSGAD IANICNEAAL HAAREGHTSV HTLNFEYAVE RVLAGTAKKS KILSKEEQKV VAFHESGHAL VGWMLEHTEA VMKVSITPRT NAALGFAQML PRDQHLFTKE QLFERMCMAL GGRASEALSF NEVTSGAQDD LRKVTRIAYS MVKQFGMAPG IGPISFPEAQ EGLMGIGRRP FSQGLQQMMD HEARLLVAKA YRHTEKVLQD NLDKLQALAN ALLEKEVINY EDIEALIGPP PHGPKKMIAP QRWIDAQREK QDLGEEETEE TQQPPLGGEE PTWPK
This gene encodes a mitochondrial metalloprotease protein that is a member of the AAA family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Mutations in this gene cause autosomal recessive spastic paraplegia 7. Two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell matrix adhesion regulator; Mitochondrial inner membrane m-AAA protease component paraplegin; Paraplegin; paraplegin, isoform 1; spastic paraplegia 7 (pure and complicated autosomal recessive); Spastic paraplegia 7 protein
Gene Aliases: CAR; CMAR; PGN; SPG5C; SPG7
UniProt ID: (Human) Q9UQ90
Entrez Gene ID: (Human) 6687
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support