Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MEGFPLVPPP SEDLVPYDTD LYQRQTHEYY PYLSSDGESH SDHYWDFHPH HVHSEFESFA ENNFTELQSV QPPQLQQLYR HMELEQMHVL DTPMVPPHPS LGHQVSYLPR MCLQYPSLSP
This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 31 kDa transforming protein; 31 kDa-transforming protein; 31kDa Transforming Protein; hCG_25181; hematopoietic transcription factor PU.1; SPIA; spleen focus forming virus (SFFV) proviral integration oncogene spi1; Transcription factor PU.1; Transcription Factor spi1
Gene Aliases: OF; PU.1; SFPI1; SPI-1; SPI-A; SPI1
UniProt ID: (Human) P17947
Entrez Gene ID: (Human) 6688
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support