Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: FKKGERDGDF ICPDYYEAVC GTDGKTYDNR CALCAENAKT GSQIGVKSEG ECKSSNPEQD VCSAFRPFVR DGRLGCTREN DPVLGPDGKT HGNKCAMCAE L
Lympho-epithelial Kazal-type inhibitor (LEKTI) is a serine protease inhibitor which protects mucous epithelia against microbial attack and inflammation. LEKTI is a marker of epithelial differentiation and expresses strongly in the granular and uppermost spinous layers of the epidermis and differentiated layers of stratified epithelia. Defects in SPINK5, the gene encoding LEKTI, are the cause of Netherton syndrome, a severe autosomal recessive disorder characterized by atopic dermatitis, hayfever and other conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ISK5; LEKTI; Lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; serine peptidase inhibitor, Kazal type 5; Serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5
Gene Aliases: 2310065D10Rik; AV238912; LEKT1; LEKTI; LETK1; LETKI; NETS; NS; SPINK5; VAKT1; VAKTI
UniProt ID: (Human) Q9NQ38
Entrez Gene ID: (Human) 11005, (Rat) 361319, (Mouse) 72432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support