Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PAAPTKALP CLPPWEGVLD MFSIKRFRAR AQLVSGHSCR LVQALPTVIR SAGCIPSNIV WDLLASICPA KAKDVCVVRL CPHGARDTQN CRLLYSYLND RQRHGLASVE HMGMVLLPLP AFQPLPTRLR PLGGPGLEVT HSSLLLAVLL PKEGLPDTAG SSPWLGKVQK MVSFNSKVEK RYYQPDDRRP NVPLKGTPPP GGAWQQSQGR GSIAPRGISA WQRPPRGRGR LWPEPENWQH PGRGQWPPEP GLRQSQHPYS VAPAGHGFGR GQHFHRDSCP HQALLRHLES LATMSHQLQA LLCPQTKSSI PRPLQRLSSA LAAPEPPGPA RDSSLGPTDE AGSECPFPRK A (854-1204 aa encoded by BC114430)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 1, regulatory subunit 146; SPOC domain-containing protein 1
Gene Aliases: PPP1R146; SPOCD1
UniProt ID: (Human) Q6ZMY3
Entrez Gene ID: (Human) 90853
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support