Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEGGLGRAV CLLTGASRGF GRTLAPLLAS LLSPGSVLVL SARNDEALRQ LEAELGAERS GLRVVRVPAD LGAEAGLQQL LGALRELPRP KGLQRLLLIN NAGSLGDVSK GFVDLSDSTQ VNNYWALNLT SMLCLTSSVL KAFPDSPGLN RTVVNISSLC ALQPFKGWAL YCAGKAARDM LFQVLALEEP NVRVLNYAPG PLDTDMQQLA RETSVDPDMR KGLQELKAKG KLVDCKVSAQ KLLSLLEKDE FKSGAHVDFY DK (1-261 aa encoded by BC017310)
SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sepiapterin reductase; short chain dehydrogenase/reductase family 38C, member 1; SPR
Gene Aliases: AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; SPR
UniProt ID: (Human) P35270, (Rat) P18297
Entrez Gene ID: (Human) 6697, (Mouse) 20751, (Rat) 29270
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support