Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: PTAILSLDQI KAIRGSNEYT EGPSVVKRPA PRTAPRQEKH ERTHEIIPIN
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
Sprouty-1 and -2 are membrane-anchored phosphoprotein inhibitors of growth factor signaling in endothelial cells. Four mammalian Sprouty homologues (Sprouty-1-4) have been identified. One study showed that overexpressed Sprouty-1 and -2 inhibits fibroblast growth factor and vascular endothelial growth factor-induced proliferation and differentiation by repressing pathways leading to MAP kinase activation. Sprouty proteins are also endogenous inhibitors of the Ras/MAP kinase pathway that play an important role in the remodeling of branching tissues, such as in development of the lung, kidney tubules, vascular system, and breast ducts. A recent study shows that Sprouty-1 and -2 may have tumor suppressing activity in the breast, and found that both proteins are consistently down-regulated in breast carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein sprouty homolog 1; sprouty homolog 1, antagonist of FGF signaling; sprouty, Drosophila, homolog of, 1 (antagonist of FGF signaling); Spry-1
Gene Aliases: hSPRY1; sprouty1; spry-1; SPRY1
UniProt ID: (Human) O43609, (Mouse) Q9QXV9
Entrez Gene ID: (Human) 10252, (Mouse) 24063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support