Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SPZ1 Antibodies

          Abnova

          SPZ1 Monoclonal Antibody (3D12)

          View all (3) SPZ1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SPZ1 Monoclonal Antibody (3D12)

          • Antibody Testing Data (2)
          SPZ1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SPZ1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SPZ1 Antibody (H00084654-M08) in WB

          SPZ1 monoclonal antibody (M08), clone 3D12. Western Blot analysis of SPZ1 expression in NIH/3T3. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SPZ1 Antibody in Western Blot (WB)
          SPZ1 Antibody in ELISA (ELISA)

          Product Details

          H00084654-M08

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          1 ng/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3D12

          Immunogen

          SPZ1 (NP_115956.2, 1 a.a. approximately 430 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MASSAKSAEM PTISKTLNPT PDPHQEYLDP RITIALFEIG SHSPSSWGSL PFLKNSSHQV TEQQTAQKFN NLLKEIKDIL KNMAGFEEKI TEAKELFEET NITEDVSAHK ENIRGLDKIN EMLSTNLPVS LAPEKEDNEK KQEMILETNI TEDVSAHKEN IRGLDKINEM LSTNLPVSLA PEKEDNEKKQ QMIMENQNSE NTAQVFARDL VNRLEEKKVL NETQQSQEKA KNRLNVQEET MKIRNNMEQL LQEAEHWSKQ HTELSKLIKS YQKSQKDISE TLGNNGVGFQ TQPNNEVSAK HELEEQVKKL SHDTYSLQLM AALLENECQI LQQRVEILKE LHHQKQGTLQ EKPIQINYKQ DKKNQKPSEA KKVEMYKQNK QAMKGTFWKK DRSCRSLDVC LNKKACNTQF NIHVARKALR GKMRSASSLR

          Target Information

          Spermatogenic leucine zipper protein 1 is a member of the basic helix-loop-helix (bHLH) family of transcription factors which is expressed specifically in the testis and epididymis. Studies show that SPZ1 acts downstream of mitogen-activated protein kinase (MAPK), an extracellular signal-regulated kinase 1/2 to up-regulate cell proliferation and tumorigenesis. Also, through an interaction with proliferating cell nuclear antigen (PCNA) promoter, SPZ1 induced cell proliferation concomitant with an increase in PCNA gene expression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BHLH-Zip transcription factor SPZ1; protein phosphatase 1, regulatory subunit 148; Spermatogenic leucine zipper protein 1; Spermatogenic Zip 1; testis secretory sperm-binding protein Li 232m; Testis-specific protein 1; Testis-specific protein NYD-TSP1

          View more View less

          Gene Aliases: 1700027M20Rik; AI429130; NYD-TSP1; PPP1R148; SPZ1; TSP1

          View more View less

          UniProt ID: (Human) Q9BXG8, (Mouse) Q99MY0

          View more View less

          Entrez Gene ID: (Human) 84654, (Mouse) 79401

          View more View less

          Function(s)
          DNA binding transcription factor activity, sequence-specific DNA binding protein binding
          Process(es)
          transcription, DNA-templated regulation of transcription, DNA-templated
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0