Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MASLTVKAYL LGKEDAAREI RRFSFCCSPE PEAEAEAAAG PGPCERLLSR VAALFPALRP GGFQAHYRDE DGDLVAFSSD EELTMAMSYV KDDIFRIYIK EKKECRRDHR PPCAQEAPRN MVHPNVICDG CNGPVVGTRY KCSVCPDYDL CSVCEGKGLH RGHTKLAFPS PFGHLSEGFS HSRWLRKVKH GHFGWPGWEM GPPGNWSPRP PRAGEARPGP TAESASGPSE DPSVNFLKNV GESVAAALSP LGIEVDIDVE HGGKRSRLTP VSPESSSTEE KSSSQPSSCC SDPSKPGGNV EGATQSLAEQ MRKIALESEG RPEEQMESDN CSGGDDDWTH LSSKEVDPST GELQSLQMPE SEGPSSLDPS QEGPTGLKEA ALYPHLPPEA DPRLIESLSQ MLSMGFSDEG GWLTRLLQTK NYDIGAALDT IQYSKHPPPL
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; oxidative stress induced; oxidative stress induced like; p62; phosphotyrosine independent ligand for the Lck SH2 domain p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; Sequestosome-1; STONE14; Ubiquitin-binding protein p62
Gene Aliases: A170; FTDALS3; ORCA; OSF-6; Osi; OSIL; p60; p62; p62B; PDB3; SQSTM1; STAP; STONE14; ZIP3
UniProt ID: (Human) Q13501, (Mouse) Q64337
Entrez Gene ID: (Human) 8878, (Mouse) 18412
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support