Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLPTQAGAA AALGRGSALG GSLNRTPTGR PGGGGGTRGA NGGRVPGNGA GLGPGRLERE AAAAAATTPA PTAGALYSGS EGDSESGEEE ELGAERRGLK RSLSEMEIGM VVGGPEASAA ATGGYGPVSG AVSGAKPGKK TRGRVKIKME FIDNKLRRYT TFSKRKTGIM KKAYELSTLT GTQVLLLVAS ETGHVYTFAT RKLQPMITSE TGKALIQTCL NSPDSPPRSD PTTDQRMSAT GFEETDLTYQ VSESDSSGET KDTLKPAFTV TNLPGTTSTI QTAPSTSTTM QVSSGPSFPI TNYLAPVSAS VSPSAVSSAN GTVLKSTGSG PVSSGGLMQL PTSFTLMPG (1-348 aa encoded by BC048211 )
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-fos serum response element-binding transcription factor; minichromosome maintenance 1 homolog; OTTHUMP00000039820; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); SRF
Gene Aliases: AW049942; AW240594; MCM1; RGD1559787; SRF
UniProt ID: (Human) P11831, (Mouse) Q9JM73
Entrez Gene ID: (Human) 6722, (Mouse) 20807, (Rat) 501099
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support