Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPFLRRRLA FLSFFWDKIW PAGGEPDHGT PGSLDPNTDP VPTLPAEPCS PFPQLFLALY DFTARCGGEL SVRRGDRLCA LEEGGGYIFA RRLSGQPSAG LVPITHVAKA SPETLSDQPW YFSGVSRTQA QQLLLSPPNE PGAFLIRPSE SSLGGYSLSV RAQAKVCHYR VSMAADGSLY LQKGRLFPGL EELLTYYKAN WKLIQNPLLQ PCMPQKAPRQ DVWERPHSEF ALGRKLGEGY FGEVWEGLWL GSLPVAIKVI KSANMKLTDL AKEIQTLKGL RHERLIRLHA VCSGGEPVYI VTELMRKGNL QAFLGTPEGR ALRLPPLLGF ACQVAEGMSY LEEQRVVHRD LAARNVLVDD GLACKVADFG LARLLKDDIY SPSSSSKIPV KWTAPEAANY RVFSQKSDVW SFGVLLHEVF TYGQCPYEGM TNHETLQQIM RGYRLPRPAA CPAEVYVLML ECWRSSPEER PSFATLREKL HAIHRCHP
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tyrosine-protein kinase Srms
Gene Aliases: C20orf148; dJ697K14.1; PTK70; SRM; SRMS
UniProt ID: (Human) Q9H3Y6
Entrez Gene ID: (Human) 6725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support