Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SSH3 Antibodies

          Abnova

          SSH3 Polyclonal Antibody, MaxPab™

          View all (13) SSH3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SSH3 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (3)
          SSH3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SSH3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SSH3 Antibody (H00054961-D01P) in WB

          Western Blot analysis of SSH3 expression in transfected 293T cell line (H00054961-T02) by SSH3 MaxPab polyclonal antibody.

          Lane 1: SSH3 transfected lysate(73.00 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SSH3 Antibody in Western Blot (WB)
          SSH3 Antibody in Western Blot (WB)
          SSH3 Antibody in Western Blot (WB)

          Product Details

          H00054961-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SSH3 (AAH07709.1, 1 a.a. approximately 659 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MALVTVSRSP PGSGASTPVG PWDQAVQRRS RLQRRQSFAV LRGAVLGLQD GGDNDDAAEA SSEPTEKAPS EEELHGDQTD FGQGSQSPQK QEEQRQHLHL MVQLLRPQDD IRLAAQLEAP RPPRLRYLLV VSTREGEGLS QDETVLLGVD FPDSSSPSCT LGLVLPLWSD TQVYLDGDGG FSVTSGGQSR IFKPISIQTM WATLQVLHQA CEAALGSGLV PGGSALTWAS HYQERLNSEQ SCLNEWTAMA DLESLRPPSA EPGGSSEQEQ MEQAIRAELW KVLDVSDLES VTSKEIRQAL ELRLGLPLQQ YRDFIDNQML LLVAQRDRAS RIFPHLYLGS EWNAANLEEL QRNRVTHILN MAREIDNFYP ERFTYHNVRL WDEESAQLLP HWKETHRFIE AARAQGTHVL VHCKMGVSRS AATVLAYAMK QYECSLEQAL RHVQELRPIA RPNPGFLRQL QIYQGILTAS RQSHVWEQKV GGVSPEEHPA PEVSTPFPPL PPEPEGGGEE KVVGMEESQA APKEEPGPRP RINLRGVMRS ISLLEPSLEL ESTSETSDMP EVFSSHESSH EEPLQPFPQL ARTKGGQQVD RGPQPALKSR QSVVTLQGSA VVANRTQAFQ EQEQGQGQGQ GEPCISSTPR FRKVVRQASV HDSGEEGEA

          Target Information

          The ADF (actin-depolymerizing factor)/cofilin family is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hSSH-3L; Protein phosphatase Slingshot homolog 3; slingshot 3; slingshot homolog 3; slingshot-3L; slingshot-like 3; SSH-3L; SSH-like protein 3

          View more View less

          Gene Aliases: BC028922; mSSH-3L; SSH-3; SSH3; SSH3L

          View more View less

          UniProt ID: (Human) Q8TE77, (Mouse) Q8K330

          View more View less

          Entrez Gene ID: (Human) 54961, (Mouse) 245857

          View more View less

          Function(s)
          DNA binding actin binding protein tyrosine phosphatase activity protein tyrosine/serine/threonine phosphatase activity phosphoprotein phosphatase activity hydrolase activity phosphatase activity
          Process(es)
          protein dephosphorylation regulation of actin polymerization or depolymerization regulation of lamellipodium assembly dephosphorylation regulation of axonogenesis peptidyl-tyrosine dephosphorylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0