Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPTMRLLSFV VLALFAVTQA EEGARLLASK SLLNRYAVEG RDLTLQYNIY NVGSSAALDV ELSDDSFPPE DFGIVSGMLN VKWDRIAPAS NVSHTVVLRP LKAGYFNFTS ATITYLAQED GPVVIGSTSA PGQGGILAQR EFDRRFSPHF LDWAAFGVMT LPSIGIPLLL WYSSKRKYDT PKTKKN
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2).The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The human beta-signal sequence receptor gene (SSR2) maps to chromosome bands 1q21-q23.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686F19123; Signal sequence receptor subunit beta; signal sequence receptor, beta (translocon-associated protein beta); SSR-beta; translocon-associated protein beta; Translocon-associated protein subunit beta; TRAP-beta
Gene Aliases: HSD25; SSR2; TLAP; TRAP-BETA; TRAPB
UniProt ID: (Human) P43308
Entrez Gene ID: (Human) 6746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support