Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MALNGAEVDD FSWEPPTEAE TKVLQARRER QDRISRLMGD YLLRGYRVLG ETCADCGTIL LQDKQRKIYC VACQELDSDV DKDNPALNAQ AALSQAREHQ LASASELPLG SRPAPQPPVP RPEHCEGAAA GLKAAQGPPA PAVPPNTDVM ACTQTALLQK LTWASAELGS STSLETSIQL CGLIRACAEA LRSLQQLQH
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autoantigen p27; centromeric autoantigen (27kD); Protein zinc ribbon domain type 2; Protein ZNRD2; Sjoegren syndrome/scleroderma autoantigen 1; Sjogren's syndrome/scleroderma autoantigen 1; Zinc ribbon domain-containing protein 2
Gene Aliases: p27; SSSCA1; ZNRD2
UniProt ID: (Human) O60232
Entrez Gene ID: (Human) 10534
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support