Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNGDDTFAK RPRDDAKASE KRSKAFDDIA TYFSKKEWKK MKYSEKISYV YMKRNYKAMT KLGFKVTLPP FMCNKQATDF QGNDFDNDHN RRIQVEHPQM TFGRLHRIIP KIMPKKPAED ENDSKGVSEA SGPQNDGKQL HPPGKANISE KINKRSGPKR GKHAWTHRLR ERKQLVIYEE ISDPEEDDE (1-188 aa encoded by BC001003 )
Could act as a modulator of transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 5.1; cancer/testis antigen family 5, member 1; CT5.1; CT5.2; CT51; Protein SSX1; sarcoma, synovial, X-chromosome-related 1; SSX2; SSX2A; SSX2B; SSX3; Synovial sarcoma, X breakpoint 1; X breakpoint 2
Gene Aliases: CT5.1; SSRC; SSX1
UniProt ID: (Human) Q16384
Entrez Gene ID: (Human) 6756
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support