Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ST7 Antibodies

          Proteintech

          ST7 Polyclonal Antibody

          1 Reference
          View all (4) ST7 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ST7 Polyclonal Antibody

          • Antibody Testing Data (16)
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ST7 Antibody (11945-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human liver cancer tissue slide using (Product # 11945-1-AP) (ST7 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Western Blot (WB)
          ST7 Antibody in Immunoprecipitation (IP)

          Product Details

          11945-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ST7 Fusion Protein Ag2547 (101-481 aa encoded by BC030954)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: LRPLLGGVD NNSSNNSDSN RQSVSECKVW RNPLNLFRGA EYNRYTWVTG REPLTYYDMN LSAQDHQTFF TCDSDHLRPA DAIMQKAWRE RNPQARISAA HEALEINECA TAYILLAEEE ATTIAEAEKL FKQALKAGDG CYRRSQQLQH HGSQYEAQHR RDTNVLVYIK RRLAMCARRL GRTREAVKMM RDLMKEFPLL SMFNIHENLL EALLELQAYA DVQAVLAKYD DISLPKSATI CYTAALLKAR AVSDKFSPEA ASRRGLSTAE MNAVEAIHRA VEFNPHVPKY LLEMKSLILP PEHILKRGDS EAIAYAFFHL AHWKRVEGAL NLLHCTWEGT FRMIPYPLEK GHLFYPYPIC TETADRELLP SFHEVSVYPK KE (101-481 aa encoded by BC030954)

          Target Information

          The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 4low density lipoprotein-related protein 12; family with sequence similarity 4, subfamily A, member 1; family with sequence similarity 4, subfamily A, member 2; mRay; Protein FAM4A1; Protein HELG; suppression of tumorigenicity 7 (breast); Suppressor of tumorigenicity 7 protein

          View more View less

          Gene Aliases: 9430001H04Rik; ETS7q; FAM4A; FAM4A1; Fam4a2; HELG; RAY1; SEN4; ST7; TSG7

          View more View less

          UniProt ID: (Human) Q9NRC1, (Mouse) Q99M96

          View more View less

          Entrez Gene ID: (Human) 7982, (Mouse) 64213

          View more View less

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0