Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IVDMHAAAD IFKHYMKYYN DYGDIIKETL SKTRQIDKIQ CAKTLILSLQ QLFNELVQEQ GPNLDRTSAH VSGIKELARR FALTFGLDQI KTREAVATLH KDGIEFAFKY QNQKGQEYPP PNLAFLEVLS EFSSKLLRQD KKTVHSYLEK FLTEQMMERR EDVWLPLISY RNSLVTGGED DRMSVNSGSS SSKTSSVRNK KGRPPLHKKR VEDESLDNTW LNRTDTMIQT PGPLPAPQLT STVLRENSRP MGDQIQEPES EHGSEPDFLH NRGLMEEDAE PIFEDVMMSS RSQLEDMNEE FEDTMVIDLP PSRNRRERAE LRPDFFDSAA IIEDDSGFGM PMF (880-1221 aa encoded by B C064699)
This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cohesin subunit SA-1; DKFZp781D1416; nuclear protein stromal antigen 1; SCC3 homolog 1; Stromal antigen 1
Gene Aliases: AU045003; SA-1; SA1; SCC3; SCC3A; STAG1
UniProt ID: (Human) Q8WVM7, (Mouse) Q9D3E6
Entrez Gene ID: (Human) 10274, (Mouse) 20842, (Rat) 315958
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support