Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MPLFTANPFE QDVEKATNEY NTTEDWSLIM DICDKVGSTP NGAKDCLKAI MKRVNHKVPH VALQALTLLG ACVANCGKIF HLEVCSRDFA TEVRAVIKNK AHPKVCEKLK SLMVEWSEEF QKDPQFSLIS ATIKSMKEEG ITFPPAGSQT VSAAAKNGTS SNKNKEDEDI AKAIELSLQE QKQQHTETKS LYPSSEIQLN NKVARKVRAL YDFEAVEDNE LTFKHGEIII VLDDSDANWW KGENHRGIGL FPSNFVTTNL NIETEAAAVD KLNVIDDDVE EIKKSEPEPV YIDEDKMDRA LQVLQSIDPT DSKPDSQDLL DLEDICQQMG PMIDEKLEEI DRKHSELSEL NVKVLEALEL YNKLVNEAPV YSVYSKLHPP AHYPPASSGV PMQTYPVQSH GGNYMGQSIH QVTVAQSYSL GPDQIGPLRS LPPNVNSSVT AQPAQTSYLS TGQDTVSNPT YMNQNSNLQS ATGTTAYTQQ MGMSVDMSSY QNTTSNLPQL AGFPVTVPAH PVAQQHTNYH QQPLL
The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp564C047; HGNC:11358; Hrs-binding protein; HSE1 homolog; Signal transducing adapter molecule 2; signal transducing adaptor molecule (SH3 domain and ITAM motif) 2; STAM-2; STAM-like protein containing SH3 and ITAM domains 2
Gene Aliases: HBP; STAM2; STAM2A; STAM2B
UniProt ID: (Human) O75886
Entrez Gene ID: (Human) 10254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support