Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • STK38 Antibodies

          Abnova

          STK38 Polyclonal Antibody, MaxPab™

          View all (15) STK38 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite STK38 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (3)
          STK38 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          STK38 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          STK38 Antibody (H00011329-D01P) in WB

          Western Blot analysis of STK38 expression in transfected 293T cell line (H00011329-T01) by STK38 MaxPab polyclonal antibody.

          Lane 1: STK38 transfected lysate(54.20 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          STK38 Antibody in Western Blot (WB)
          STK38 Antibody in Western Blot (WB)
          STK38 Antibody in Western Blot (WB)

          Product Details

          H00011329-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          STK38 (NP_009202.1, 1 a.a. approximately 465 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAMTGSTPCS SMSNHTKERV TMTKVTLENF YSNLIAQHEE REMRQKKLEK VMEEEGLKDE EKRLRRSAHA RKETEFLRLK RTRLGLEDFE SLKVIGRGAF GEVRLVQKKD TGHVYAMKIL RKADMLEKEQ VGHIRAERDI LVEADSLWVV KMFYSFQDKL NLYLIMEFLP GGDMMTLLMK KDTLTEEETQ FYIAETVLAI DSIHQLGFIH RDIKPDNLLL DSKGHVKLSD FGLCTGLKKA HRTEFYRNLN HSLPSDFTFQ NMNSKRKAET WKRNRRQLAF STVGTPDYIA PEVFMQTGYN KLCDWWSLGV IMYEMLIGYP PFCSETPQET YKKVMNWKET LTFPPEVPIS EKAKDLILRF CCEWEHRIGA PGVEEIKSNS FFEGVDWEHI RERPAAISIE IKSIDDTSNF DEFPESDILK PTVATSNHPE TDYKNKDWVF INYTYKRFEG LTARGAIPSY MKAAK

          Target Information

          STK38 (also known as NDR) or Nuclear Dbf2-related protein is a serine/threonine kinase in the subgroup of the AGC kinase family that is regulated by Mob proteins. Fas and TNFα receptor stimulation activates human STK38 by promoting phosphorylation at the hydrophobic motif. NDR acts as a novel pro-apoptotic kinase, and is a key member of the RASSF1A/MST1 signaling cascade. STK38 is crucial for the fidelity of mitotic chromosome alignment in mammalian cells. STK38 is incorporated into the HIV-1 particles and is cleaved by the HIV-1 protease. STK38-driven centrosome duplication requires Cdk2 activity, and Cdk2-induced centrosome amplification is affected upon reduction of STK38 activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Ndr Ser/Thr kinase-like protein; NDR1 protein kinase; Nuclear Dbf2-related kinase 1; Serine/threonine-protein kinase 38; stk38

          View more View less

          Gene Aliases: 5830476G13Rik; 9530097A09Rik; AA617404; NDR; NDR1; STK38

          View more View less

          UniProt ID: (Human) Q15208, (Mouse) Q91VJ4

          View more View less

          Entrez Gene ID: (Human) 11329, (Mouse) 106504

          View more View less

          Function(s)
          magnesium ion binding protein serine/threonine kinase activity protein binding ATP binding mitogen-activated protein kinase kinase kinase binding cadherin binding involved in cell-cell adhesion nucleotide binding protein kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups metal ion binding
          Process(es)
          cellular protein modification process protein phosphorylation phosphorylation intracellular signal transduction negative regulation of MAP kinase activity peptidyl-serine phosphorylation cell-cell adhesion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0