Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EHRIGAPGVE EIKSNSFFEG VDWEHIRERP AAISIEIKSI DDTSNFDEFP ESDILKPTVA TSNHPETDYK NKDWVFINYT YKRFEGLTAR GAIPSYMKAA K
STK38 (also known as NDR) or Nuclear Dbf2-related protein is a serine/threonine kinase in the subgroup of the AGC kinase family that is regulated by Mob proteins. Fas and TNFα receptor stimulation activates human STK38 by promoting phosphorylation at the hydrophobic motif. NDR acts as a novel pro-apoptotic kinase, and is a key member of the RASSF1A/MST1 signaling cascade. STK38 is crucial for the fidelity of mitotic chromosome alignment in mammalian cells. STK38 is incorporated into the HIV-1 particles and is cleaved by the HIV-1 protease. STK38-driven centrosome duplication requires Cdk2 activity, and Cdk2-induced centrosome amplification is affected upon reduction of STK38 activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ndr Ser/Thr kinase-like protein; NDR1 protein kinase; Nuclear Dbf2-related kinase 1; Serine/threonine-protein kinase 38
Gene Aliases: NDR; NDR1; STK38
UniProt ID: (Human) Q15208
Entrez Gene ID: (Human) 11329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support