Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: CNIPPQEIL TRDSVTTQVD GVVYYRIYSA VSAVANVNDV HQATFLLAQT TLRNVLGTQT LSQILAGREE IAHSIQTLLD DATELWGIRV ARVEIKDVRI PVQLQRSMAA EAEATREARA KVLAAEGEMN ASKSLKSASM VLAESPIALQ LRYLQTLSTV ATEKNSTIVF PLPMNILEGI GGVSYDNHKK LPNKA (98-291 aa encoded by B C025760)
STOML3 is required for the function of many mechanoreceptors. It modulates mechanotransduction channels and acid-sensing ion channels (ASIC) proteins. The protein potentiates PIEZO1 and PIEZO2 function by increasing their sensitivity to mechanical stimulations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EPB72-like 3; erythrocyte band 7 integral membrane protein; protein 7.2B; SLP-3; stomatin (EPB72)-like 3; stomatin related olfactory protein SRO; Stomatin-like protein 3; Stomatin-related olfactory protein
Gene Aliases: Epb7.2l; SLP3; SRO; STOML3
UniProt ID: (Human) Q8TAV4, (Mouse) Q6PE84
Entrez Gene ID: (Human) 161003, (Rat) 295041, (Mouse) 229277
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support