Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKYSCCALVL AVLGTELLGS LCSTVRSPRF RGRIQQERKN IRPNIILVLT DDQDVELGSL QVMNKTRKIM EHGGATFINA FVTTPMCCPS RSSMLTGKYV HNHNVYTNNE NCSSPSWQAM HEPRTFAVYL NNTGYRTAFF GKYLNEYNGS YIPPGWREWL GLIKNSRFYN YTVCRNGIKE KHGFDYAKDY FTDLITNESI NYFKMSKRMY PHRPVMMVIS HAAPHGPEDS APQFSKLYPN ASQHITPSYN YAPNMDKHWI MQYTGPMLPI HMEFTNILQR KRLQTLMSVD DSVERLYNML VETGELENTY IIYTADHGYH IGQFGLVKGK SMPYDFDIRV PFFIRGPSVE PGSIVPQIVL NIDLAPTILD IAGLDTPPDV DGKSVLKLLD PEKPGNRFRT NKKAKIWRDT FLVERGKFLR KKEESSKNIQ QSNHLPKYER VKELCQQARY QTACEQPGQK WQCIEDTSGK LRIHKCKGPS DLLTVRQSTR NLYARGFHDK DKECSCRESG YRASRSQRKS QRQFLRNQGT PKYKPRFVHT RQTRSLSVEF EGEIYDINLE EEEELQVLQP RNIAKRHDEG HKGPRDLQAS SGGNRGRMLA DSSNAVGPPT TVRVTHKCFI LPNDSIHCER ELYQSARAWK DHKAYIDKEI EALQDKIKNL REVRGHLKRR KPEECSCSKQ SYYNKEKGVK KQEKLKSHLH PFKEAAQEVD SKLQLFKENN RRRKKERKEK RRQRKGEECS LPGLTCFTHD NNHWQTAPFW NLGSFCACTS SNNNTYWCLR TVNETHNFLF CEFATGFLEY FDMNTDPYQL TNTVHTVERG ILNQLHVQLM ELRSCQGYKQ CNPRPKNLDV GNKDGGSYDL HRGQLWDGWE G
Heparan sulfate proteoglycans act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arylsulfatase; Extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; N-acetylglucosamine-6-sulfatase; sulfatase FP
Gene Aliases: KIAA1077; SULF-1; SULF1
UniProt ID: (Human) Q8IWU6
Entrez Gene ID: (Human) 23213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support