Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SYCE1 (synaptonemal complex central element protein 1), also known as CT76 (cancer/testis antigen 76), is a 351 amino acid nuclear protein that belongs to the SYCE family and exists as three alternatively spliced isoforms. Colocalizing with SYCE2 on chromatin, SYCE1 exists as a homodimer and is found in the transverse central elements of the central region between lateral elements of the synaptonemal complexes (SCS). The transverse central element of the SCS is formed between homologous chromosomes during meiotic prophase and requires SYCP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 76; CT76; Synaptonemal complex central element protein 1; telomeric protein
Gene Aliases: 4933406J07Rik; C10orf94; CT76; POF12; SPGF15; SYCE1
UniProt ID: (Human) Q8N0S2, (Mouse) Q9D495, (Rat) Q5XHZ2
Entrez Gene ID: (Human) 93426, (Mouse) 74075, (Rat) 502372
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support