Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNGRVDYLVT EEEINLTRGP SGLGFNIVGG TDQQYVSNDS GIYVSRIKEN GAAALDGRLQ EGDKILSVNG QDLKNLLHQD AVDLFRNAGY AVSLRVQHRL QVQNGPIGHR GEGDPSGIPI FMVLVPVFAL TMVAAWAFMR YRQQL
Regulates endocytosis of activin type 2 receptor kinases through the Ral/RALBP1-dependent pathway and may be involved in suppression of activin-induced signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin receptor interacting protein 5; Mitochondrial outer membrane protein 25; Synaptojanin-2-binding protein
Gene Aliases: ARIP2; OMP25; SYNJ2BP
UniProt ID: (Human) P57105
Entrez Gene ID: (Human) 55333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support