Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QVLWEEQDQ CLQELSREQT GDLGTEQPVP GCEGMWDNIS CWPSSVPGRM VEVECPRFLR MLTSRNGSLF RNCTQDGWSE TFPRPNLACG VNVNDSSNEK RHSYLLKL (36-142 aa encoded by BC035757 )
The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MYOP; pancreatic secretin receptor; SCT-R; Secretin receptor
Gene Aliases: 6530402O03Rik; SCTR; SR
UniProt ID: (Human) P47872, (Rat) P23811, (Mouse) Q5FWI2
Entrez Gene ID: (Human) 6344, (Rat) 81779, (Mouse) 319229
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support