Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total SEPT10.
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11619; Septin-10
Gene Aliases: 4921515A04Rik; 9430099J10Rik; AA408298; AI874685; SEPT10; SEPTIN10
UniProt ID: (Human) Q9P0V9, (Rat) Q5PQK1, (Mouse) Q8C650
Entrez Gene ID: (Human) 151011, (Rat) 309891, (Mouse) 103080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support