Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Description: This SMMS-1 monoclonal antibody reacts with both isoforms of the human smooth muscle myosin heavy chain (SM1 and SM2) structural proteins, which are components of the contractile apparatus in smooth muscle cells. Smooth muscle myosin heavy chains are expressed by myoepithelial cells and also serve as the terminal differentiation marker for smooth muscle cells. In normal breast tissue, myoepithelial cells compose the outer layer of the lumen lining. In benign and in situ breast lesions, the intact myoepithelial layer stains with SMMS-1 and the loss of this staining indicates invasive cancer. This SMMS-1 antibody has also been reported to cross-react with bovine, canine, feline, and rat smooth muscle myosin heavy chains.
Applications Reported: This SMMS-1 antibody has been reported for use in microscopy, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.
Applications Tested: This SMMS-1 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 30 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: myh11 {ECO:0000312|RGD:3136}; Myosin heavy chain 11; myosin heavy chain 21; myosin heavy chain SMA; Myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; Myosin-11; SMMHC; smooth muscle myosin heavy chain
Gene Aliases: AAT4; BOS_22758; FAA4; KIAA0866; MYH11; SMHC; SMMHC
UniProt ID: (Human) P35749
Entrez Gene ID: (Bovine) 530050, (Human) 4629, (Dog) 479836, (Rat) 24582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support