Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SPG20 Antibodies

          Proteintech

          Spartin/SPG20 Polyclonal Antibody

          16 References
          View all (12) SPG20 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Spartin/SPG20 Polyclonal Antibody

          • Antibody Testing Data (18)
          Spartin/SPG20 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Spartin/SPG20 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Spartin/SPG20 Antibody (13791-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using SPG20 antibody 13791-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Spartin/SPG20 Antibody in Immunocytochemistry (ICC/IF)
          Spartin/SPG20 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Spartin/SPG20 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Spartin/SPG20 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Spartin/SPG20 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Spartin/SPG20 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Western Blot (WB)
          Spartin/SPG20 Antibody in Flow Cytometry (Flow)
          Spartin/SPG20 Antibody in Flow Cytometry (Flow)
          Spartin/SPG20 Antibody in Immunoprecipitation (IP)
          Spartin/SPG20 Antibody in Immunoprecipitation (IP)

          Product Details

          13791-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Spartin, SPG20 Fusion Protein Ag4815 (1-350 aa encoded by BC047083)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.15 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MEQEPQNGE PAEIKIIREA YKKAFLFVNK GLNTDELGQK EEAKNYYKQG IGHLLRGISI SSKESEHTGT GWESARQMQQ KMKETLQNVR TRLEILEKGL ATSLQNDLQE VPKLYPEFPP KDMCEKLPEP QSFSSAPQHA EVNGNTSTPS AGAVAAPASL SLPSQSCPAE APPAYTPQAA EGHYTVSYGT DSGEFSSVGE EFYRNHSQPP PLETLGLDAD ELILIPNGVQ IFFVNPAGEV SAPSYPGYLR IVRFLDNSLD TVLNRPPGFL QVCDWLYPLV PDRSPVLKCT AGAYMFPDTM LQAAGCFVGV VLSSELPEDD RELFEDLLRQ MSDLRLQANW NRAEEENEFQ I (1-350 aa encoded by BC047083)

          Target Information

          SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Spartin; spastic paraplegia 20 (Troyer syndrome) homolog; Spastic paraplegia 20 protein; spastic paraplegia 20, spartin (Troyer syndrome) homolog; Trans-activated by hepatitis C virus core protein 1

          View more View less

          Gene Aliases: AI840044; C79168; KIAA0610; mKIAA0610; SPART; SPARTIN; SPG20; TAHCCP1

          View more View less

          UniProt ID: (Human) Q8N0X7, (Mouse) Q8R1X6

          View more View less

          Entrez Gene ID: (Human) 23111, (Rat) 295053, (Mouse) 229285

          View more View less

          Function(s)
          protein binding ubiquitin protein ligase binding
          Process(es)
          abscission negative regulation of BMP signaling pathway lipid particle organization negative regulation of collateral sprouting in absence of injury neuromuscular process cell division regulation of mitochondrial membrane potential adipose tissue development cytokinesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0