Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVWIFQAYL AFQDLRNDAP VPYKRFLDEG GMVLTTLPLP SANSPVNMPT TGPNSLSYAS SALSPCLTAP KSPRLAMMPD N (155-234 aa encoded by BC106891)
Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. SYNGR3 is predominantly expressed in brain and placenta. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC125805; synaptogyrin 4 protein; Synaptogyrin-4; testis tissue sperm-binding protein Li 72n
Gene Aliases: 1700016O14Rik; SYNGR4
UniProt ID: (Human) O95473, (Mouse) Q9Z1L2
Entrez Gene ID: (Human) 23546, (Mouse) 58867, (Rat) 292916
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support